Orders LinkContactUsLinkComponent
Disposable tip adapters
Disposable tip adapters
Disposable tip adapter, CTFE, 100 µl
Katalog # HAMI31330
Leverandør:   HAMILTON BONADUZ
Disposable tip adapters
Katalog # HAMI31330
Leverandør:  HAMILTON BONADUZ
Disposable tip adapters
Disposable tip adapters
Disposable tip adapter, CTFE, 100 µl
Price
341,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
These adapters enable the connection of a standard 1 ml or 200 µl disposable pipette tip to the end of a Hamilton syringe. The adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes. When used in conjunction the PB600-1 and the disposable tip adapters can be used as a small volume repeating pipette.
  • Ideal for eliminating the possibility for cross contamination
Specifikationer
  • Beskrivelse:
    Disposable tip adapter, CTFE, 100 µl
  • Seq:
    01