Disposable tip adapters
Disposable tip adapter, CTFE, 100 µl
Katalog # HAMI31330
Leverandør: HAMILTON BONADUZ
Gå til:
Product Details & Documents
These adapters enable the connection of a standard 1 ml or 200 µl disposable pipette tip to the end of a Hamilton syringe. The adapter connects to standard Luer Tip (LT) and PTFE Luer Lock (TLL) syringes. When used in conjunction the PB600-1 and the disposable tip adapters can be used as a small volume repeating pipette.
- Ideal for eliminating the possibility for cross contamination
Specifikationer
- Beskrivelse:Disposable tip adapter, CTFE, 100 µl
- Seq:01