Orders LinkContactUsLinkComponent
Electrolyte Solutions
Electrolyte Solutions
Elektrolyt Skylyte, 100 ml, 100 ml
Katalog # 662-9203
Leverandør:   HAMILTON BONADUZ
Electrolyte Solutions
Katalog # 662-9203
Leverandør:  HAMILTON BONADUZ
CAS nummer:  
Electrolyte Solutions
Electrolyte Solutions
Elektrolyt Skylyte, 100 ml, 100 ml
CAS nummer:  
Price
308,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
A variety of electrolyte solutions for pH and ORP electrodes.
Specifikationer
Specifikationer
  • Pakn:
    100 ml
  • Beskrivelse:
    Elektrolyt Skylyte, 100 ml