Syringe guides
Syringe guide for Series 702 through 750 Microliter™ syringes, 7000 Modified Microliter™ syringes and series 1702 through 1750 Gastight® syringes
Catalog # HAMI14906
Supplier: HAMILTON BONADUZ
Jump to:
Product Details & Documents
The syringe guide is a plunger support device. It is compatible with the 700, 1700 and 7000 series syringes. With only two guide arms the syringe guide protects the plunger from damage but should not be used to improve accuracy and precision.
Specifications
- Beskrivelse:Syringe guide for Series 702 through 750 Microliter™ syringes, 7000 Modified Microliter™ syringes and series 1702 through 1750 Gastight® syringes
- Seq:01