Orders LinkContactUsLinkComponent
Syringe guides
Syringe guides
Syringe guide for Series 701 Microliter™ and 1701 Gastight® syringes
Katalog # HAMI14806
Leverandør:   HAMILTON BONADUZ
Syringe guides
Katalog # HAMI14806
Leverandør:  HAMILTON BONADUZ
Syringe guides
Syringe guides
Syringe guide for Series 701 Microliter™ and 1701 Gastight® syringes
Price
474,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
The syringe guide is a plunger support device. It is compatible with the 700, 1700 and 7000 series syringes. With only two guide arms the syringe guide protects the plunger from damage but should not be used to improve accuracy and precision.
Specifikationer
  • Beskrivelse:
    Syringe guide for Series 701 Microliter™ and 1701 Gastight® syringes
  • Seq:
    02