Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Incinerating dishes, rectagular

50 mm, 36 mm, 38 mm
  HALDAL/50X38X36
Incinerating dishes, rectagular
  HALDAL/50X38X36
Incinerating dishes, rectagular
Incinerating dishes, rectagular

50 mm, 36 mm, 38 mm

 
883,00 kr
   
1 * 1 Items
 

Alsint 99,7.

  • Good thermal shock resistance due to high thermal conductivity
  • Resistant to chemical attack, e.g. hydrogen and other reducing gases
 
  • Bredde:
    38 mm
  • Højde:
    36 mm
  • Længde:
    50 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.