Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Molybdate reagent

Molybdate reagent

Molybdate reagent for phosphate determination, 100 ml, 100 mL
Katalog # HACH223632
Leverandør:   Hach
Molybdate reagent
Katalog # HACH223632
Leverandør:  Hach
CAS nummer:  
Molybdate reagent
Molybdate reagent

Molybdate reagent for phosphate determination, 100 ml, 100 mL

CAS nummer:  
Price
271,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

Determination of high range phosphate (phosphorus) by the amino acid method.

Documents

Specifikationer

Specifikationer

  • Pakn:
    100 mL
  • Beskrivelse:
    Molybdate reagent for phosphate determination, 100 ml

Specifikationer