Orders LinkContactUsLinkComponent
Aluminium standard solution
Aluminium standard solution
Aluminium, Al, 100 mg/L, 100 mL
Katalog # HACH1417442
Leverandør:   Hach
Aluminium standard solution
Katalog # HACH1417442
Leverandør:  Hach
CAS nummer:  
Aluminium standard solution
Aluminium standard solution
Aluminium, Al, 100 mg/L, 100 mL
CAS nummer:  
Price
353,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
Standard solution for accuracy checks in aluminium determinations.
Documents
Specifikationer
Specifikationer
  • Koncentration:
    100 mg/L
  • Pakn:
    100 mL
  • Element:
    Aluminium
  • Symbol:
    Al
Specifikationer