Orders LinkContactUsLinkComponent
Colour standards
Colour standards
Standard colour solution, 500 Pt-Co units, Bottle, 1× 1 litre, 1 L
Katalog # 707-0002
Leverandør:   Hach
Colour standards
Katalog # 707-0002
Leverandør:  Hach
CAS nummer:  
Colour standards
Colour standards
Standard colour solution, 500 Pt-Co units, Bottle, 1× 1 litre, 1 L
CAS nummer:  
Price
3.830,00 kr
Log ind for tilgængelighed
1 * 1 L
Product Details & Documents
For accuracy checks in colour determinations.
  • Simple Accuracy Checks
  • Minimize waste with Voluette ampoules
  • Reliable results
The voluette format has the following advantages: Open a fresh standard amopule for each accuracy check and each ampule contains enough standard for one set of standard additions. Sealed ampoules prevent accidental contamination or evaporation of standard solutions.
Specifikationer
Specifikationer
  • Pakning:
    Bottle
  • Pakn:
    1 L
  • Beskrivelse:
    Standard colour solution, 500 Pt-Co units
  • Pk Info:
    1× 1 litre