Orders LinkContactUsLinkComponent
Inoculating loops and needles, disposable
Nåle, -, gul, 50 nåle pr. hylster, 40 hylstre pr. æske
  391-0249
Inoculating loops and needles, disposable
  391-0249
Inoculating loops and needles, disposable
Inoculating loops and needles, disposable
Nåle, -, gul, 50 nåle pr. hylster, 40 hylstre pr. æske
1.050,00 kr
1 * 2.000 Items
Flexible polymer.
  • Sterilised by irradiation
  • Individually wrapped
  • Length: 200 mm
  • Pakket:
    50 nåle pr. hylster, 40 hylstre pr. æske
  • Kapacitet:
    -
  • Farve:
    gul
  • Beskrivelse:
    Nåle
  • Seq:
    08