Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Autoclavable Bags

Autoclavable Bags

PA, 130 l, 1100×700 mm
Katalog # 129-9724
Leverandør:   Greiner Bio-One
Autoclavable Bags
Katalog # 129-9724
Leverandør:  Greiner Bio-One
Autoclavable Bags
Autoclavable Bags

PA, 130 l, 1100×700 mm

Price
5.010,00 kr
Log ind for tilgængelighed
1 * 200 Items
Product Details & Documents

These bags are designed specifically for the disposal of contaminated laboratory waste, that first requires steam sterilisation.

  • Bags for autoclaving and oven - resistant up to 160 ° C in an oven and 134 ° C in an autoclave
  • Suitable for hot air sterilisers
Specifikationer
  • Kapacitet:
    130 l
  • L×B:
    1100×700 mm
  • Materiale:
    PA