Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Autoclavable Bags

Autoclavable Bags

PA, 65 l, 780×600 mm
Katalog # 129-9723
Leverandør:   Greiner Bio-One
Autoclavable Bags
Katalog # 129-9723
Leverandør:  Greiner Bio-One
Autoclavable Bags
Autoclavable Bags

PA, 65 l, 780×600 mm

Price
4.830,00 kr
Log ind for tilgængelighed
1 * 300 Items
Product Details & Documents

These bags are designed specifically for the disposal of contaminated laboratory waste, that first requires steam sterilisation.

  • Bags for autoclaving and oven - resistant up to 160 ° C in an oven and 134 ° C in an autoclave
  • Suitable for hot air sterilisers
Specifikationer
  • Kapacitet:
    65 l
  • L×B:
    780×600 mm
  • Materiale:
    PA
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.