Centrifugerør
4,5 ml, 12,4×75 mm, Ventileret, transparent, Round bottom, sterile, Pakke med 25
Katalog # 212-3166
Leverandør: Greiner Bio-One
Gå til:
Product Details & Documents
Transparent PS or PP, 2-position ventilation stopper, round bottom, sterile.
- PS tubes without graduations, good thermal, mechanical and chemical resistance, centrifugation RFC 5000×g (fix-angle rotor) or 4800×g (swinging-bucket rotor)
- PP tubes with graduations; high thermal, mechanical and chemical resistance, centrifugation RCF 34000×g (fix-angle rotor) or 5800×g (swinging-bucket rotor)
- The ventilation stopper has two functions: when pushed in gently, it provides uniform ventilation of the tube contents; when pushed in firmly, it provides a tight seal
- Free from DNase/RNase and human DNA, non-pyrogenic
: Available in packs of 25 or wrapped individually.
Certifikater
Søg efter partinummerSpecifikationer
- Lukkestørrelse:Ventileret
- Ø×L:12,4×75 mm
- Pakket:Pakke med 25
- Model:Round bottom
- Version:sterile
- Kapacitet:4,5 ml
- Lågfarve:transparent