Orders LinkContactUsLinkComponent
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
50 ml, 12/21
Katalog # POUL1.51545F
Leverandør:   Poulten & Graf
Volumetric flasks, class A, white markings
Katalog # POUL1.51545F
Leverandør:  Poulten & Graf
Volumetric flasks, class A, white markings
Volumetric flasks, class A, white markings
50 ml, 12/21
Price
283,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass, amber, with PP stopper, conformity certified.
  • With ground ring mark
  • White markings
Specifikationer
  • NS:
    12/21
  • Kapacitet:
    50 ml