Volumetric flasks, class A, white markings
25 ml, 10/19
POUL1.51543F
Borosilicate glass, amber, with PP stopper, conformity certified.
- With ground ring mark
- White markings
- NS:10/19
- Kapacitet:25 ml