Orders LinkContactUsLinkComponent
Hydrometers
1,300 - 1,400 g/ml, 0,001 g/ml
Catalog # GERI15/1.3-1.4
Supplier:   GERING
Hydrometers
Catalog # GERI15/1.3-1.4
Supplier:  GERING
Hydrometers
Hydrometers
1,300 - 1,400 g/ml, 0,001 g/ml
Price
476,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Seq:
    08
  • Inddeling:
    0,001 g/ml
  • Område:
    1,300 - 1,400 g/ml