Orders LinkContactUsLinkComponent
Density hydrometers, M100
Density hydrometers, M100
M100, 1,100 - 1,200 g/ml, 0,002 g/ml, 245 mm
Catalog # GERI0348
Supplier:   GERING
Density hydrometers, M100
Catalog # GERI0348
Supplier:  GERING
Density hydrometers, M100
Density hydrometers, M100
M100, 1,100 - 1,200 g/ml, 0,002 g/ml, 245 mm
Price
368,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Reference temperature for density measurements: 20 °C.
Certifications: DIN 12791
Packaging: In velvet lined case.
Specifikationer
  • Længde:
    245 mm
  • Seq:
    05
  • Inddeling:
    0,002 g/ml
  • Model:
    M100
  • Område:
    1,100 - 1,200 g/ml