Orders LinkContactUsLinkComponent
Pipette calibration software, PICASO
Moisture trap with lid
  EPPE3113809.000
   Lav fremstillingspåvirkning
 :   EPPENDORF
Pipette calibration software, PICASO
  EPPE3113809.000
 :  EPPENDORF
   Lav fremstillingspåvirkning
Pipette calibration software, PICASO
Pipette calibration software, PICASO
Moisture trap with lid
2.510,00 kr
1 * 1 Items
PICASO (PipetteCalibrationSoftware) is a program for the automatic input and processing of measuring data from the gravimetric or photometric pipette test. With PICASO, all types of pipettes and other dispensers (e.g. bottle-top dispensers) can be checked.
  • User-oriented Windows interface, multilingual
  • On-line operating instructions and help function
  • Complete storage of a measurement including all individual values
  • Transfer of the data to other Windows programs (e.g. Excel) possible
  • Supports the archiving of complete data records
  • Analytical balances (Mettler (model AT and AE), Sartorius (model RC and MC), OHAUS (model AP) and AnD (model HM)can be connected via RS232 interface
System requirements:
IBM compatible computer in following minimum configuration:
– CPU: min. 80486/40Mhz under Windows 3.1 or 3.11 or min. Intel - Pentium/75Mhz under Windows 95
– RAM: 8 MB under 3.1 Windows or 3.11; 16 MB under Windows 95
– Hard disk (EIDE or SCSI) with at least 20 MB free disk space
– VGA monitor
  • Beskrivelse:
    Moisture trap with lid
  • Cat. No.:
    EPPE3113809.000
  • Miljøcertificering:
    Renewable Energy Certificate, ISO 14001
  • Environmentally Preferable:
    Lav fremstillingspåvirkning