Testsigter, 200×50 mm
200 mm, 50 mm, 2,00 mm
Catalog # 510-0371
Supplier: ENDECOTTS
Jump to:
Product Details & Documents
Testsigter fremstillet af rustfrit stål.
: I henhold til ASTM E 11-95
: Hver sigte leveres med individuelt kvalitetscertifikat.
Specifications
- Ø:200 mm
- Netstørrelse:2,00 mm
- Højde:50 mm
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



