Orders LinkContactUsLinkComponent
Accessories for Test Sieves
Accessories for Test Sieves
Intermediate collector, 200 mm, Rustfrit stål
Catalog # ENDE200S/STINT
Supplier:   ENDECOTTS
Accessories for Test Sieves
Catalog # ENDE200S/STINT
Supplier:  ENDECOTTS
Accessories for Test Sieves
Intermediate collector, 200 mm, Rustfrit stål
Price
1.090,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Specifications
  • Ø:
    200 mm
  • Beskrivelse:
    Intermediate collector
  • Materiale:
    Rustfrit stål
  • Seq:
    09