Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Carbon standards

Carbon standards

Mixed carbon standard, 100 ppm, 500 mL
Catalog # REGETOIC100
Supplier:   REAGECON
Carbon standards
Catalog # REGETOIC100
Supplier:  REAGECON
CAS Number:  
Carbon standards

Mixed carbon standard, 100 ppm, 500 mL

CAS Number:  
Price
2.050,00 kr
Log in for availability
1 * 500 mL
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents

These standards are manufactured from high purity raw materials and is suitable for calibrating a wide range of Total Organic Carbon (TOC) and Total Inorganic Carbon (TIC) analysers to a high accuracy.

These standards are independent, traceable and certified.

Certifications: Accredited to IEC/ISO 17025 and ISO 17034.

Documents

Specifications

Specifications

  • Koncentration:
    100 ppm
  • Pakn:
    500 mL
  • Beskrivelse:
    Mixed carbon standard

Specifications