Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
E-Set Trace calibration set, 6 bottles with 50 ml calibration and control standard each, 0,01 mol/l KCl, 1413 µS/cm at 25 °C, accuracy ± 0.5 %, traceable to PTB/NIST, 1 SET
Katalog # 663-1891
Leverandør:   WTW
Conductivity standards
Katalog # 663-1891
Leverandør:  WTW
CAS nummer:  
Conductivity standards
Conductivity standards
E-Set Trace calibration set, 6 bottles with 50 ml calibration and control standard each, 0,01 mol/l KCl, 1413 µS/cm at 25 °C, accuracy ± 0.5 %, traceable to PTB/NIST, 1 SET
CAS nummer:  
Price
1.530,00 kr
Log ind for tilgængelighed
1 * 1 Each
Specifikationer
Specifikationer
  • Pakn:
    1 SET
  • Beskrivelse:
    E-Set Trace calibration set, 6 bottles with 50 ml calibration and control standard each, 0,01 mol/l KCl, 1413 µS/cm at 25 °C, accuracy ± 0.5 %, traceable to PTB/NIST