Buffer solutions as per DIN 19266-01: 2000
Buffer solution, PL 4, 4,01, ±0,02, 250 mL
Leverandør: WTW
Gå til:
Product Details & Documents
These standard buffer solutions (work reference buffer solutions) are widely used in precision applications.
- Traceably certified to PTB/NIST and therefore safe for use
- Every bottle is labeled with their own lot number with an expiration date
Certifications: DIN 19266-01: 2000.
Documents
Certifikater
Søg efter partinummercoc_WTWA109000_07.27 CoC_WTWA109000_08.24-1 CoC_WTWA109000_12.23-1 CoC_WTWA109110_01.24-1 CoC_WTWA109110_02.27-1
More
Sensors for Field and Laboratory.pdf
Yderligere produktdetaljer
WTWA109110_TDS_EN
Datablad
Specifikationer
Specifikationer
- Pakn:250 mL
- Beskrivelse:Buffer solution, PL 4
- pH nøjagtighed:±0,02
- pH-værdi:4,01
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



