Orders LinkContactUsLinkComponent
Vial racks, WHEATON®
Vial racks, WHEATON®
30 mm, 50, 337×170×30 mm
Katalog # 548-1976
Leverandør:   DWK Life Sciences
Vial racks, WHEATON®
Katalog # 548-1976
Leverandør:  DWK Life Sciences
Vial racks, WHEATON®
Vial racks, WHEATON®
30 mm, 50, 337×170×30 mm
Price
4.690,00 kr
Log ind for tilgængelighed
1 * 5 Items
Product Details & Documents
These PP racks accommodate 48, 50, 90 or 96 vials of different sizes.
  • Autoclavable and stackable
  • Good chemical resistance
  • Includes position identifiers
Specifikationer
  • Hul Ø:
    30 mm
  • Ant. huller:
    50
  • Seq:
    02
  • B×D×H:
    337×170×30 mm