Orders LinkContactUsLinkComponent
Vial racks, WHEATON®
Vial racks, WHEATON®
23,1 mm, 36, 322×91×28 mm
Catalog # 212-6191
Supplier:   DWK Life Sciences
Vial racks, WHEATON®
Catalog # 212-6191
Supplier:  DWK Life Sciences
Vial racks, WHEATON®
Vial racks, WHEATON®
23,1 mm, 36, 322×91×28 mm
Price
2.630,00 kr
Log in for availability
1 * 5 Items
Product Details & Documents
These polypropylene racks accommodate 36 or 50 headspace auto-sampler vials. Each well has an opening in the bottom to facilitate drainage.
  • Autoclavable and stackable
  • Chemically resistant
  • Include position identifiers
Specifications
  • Hul Ø:
    23,1 mm
  • Ant. huller:
    36
  • B×D×H:
    322×91×28 mm