Trypsinising flasks, with screw caps, WHEATON®
150 ml, Topskruelåg 38-430
Katalog # WHEA355394
Leverandør: DWK Life Sciences
Gå til:
Product Details & Documents
Made of borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These trypsinising flasks units have screw caps with white styrene-butadiene rubber liners that reduce contamination of contents. Fluted design improves mixing.
Certifications: Conforms to USP Type I and ASTM E-438 Type I, Class A requirements.
Specifikationer
- Kapacitet:150 ml
- Beskrivelse:Topskruelåg 38-430