Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Aluminium seals, lined, WHEATON®

Natural, open top, PTFE/red rubber, 11 mm, aluminium
  218-2102
Aluminium seals, lined, WHEATON®
  218-2102
Aluminium seals, lined, WHEATON®
Aluminium seals, lined, WHEATON®

Natural, open top, PTFE/red rubber, 11 mm, aluminium

 
2.430,00 kr
   
1 * 1.000 Items
 

Open top seal provides for filling or retrieving contents with a syringe, large target diameter.

  • For use with 12×32 mm crimp top vials
  • Choice of liner

They are also ideal for use as closures in chromatography and instrumentation applications.

 
  • Ø:
    11 mm
  • Septum:
    PTFE/red rubber
  • Model:
    Natural, open top
  • Materiale:
    aluminium
  • Seq:
    02