Opsamlingsplader, Whatman UniPlate™
PP, 48-brønds (rektangulær), ufarvet, 5000 μl
Katalog # 734-2561
Leverandør: Whatman products (Cytiva)
Gå til:
Product Details & Documents
The UNIPLATE™ series includes microplates with a range of well profiles, well volumes and well densities, made from diverse polymer materials. Most UNIPLATE™ microplates conform to the proposed SBS microplate standard and fit most microplate readers and automated plate handling devices.
They are suitable for a wide range of applications, including simple filtrate collection, when used in conjunction with UNIFILTER® microplates, as well as homogeneous assay techniques utilised in HTS.
Specifikationer
- Farve:ufarvet
- Beskrivelse:PP, 48-brønds (rektangulær)
- Brøndvolumen:5000 μl