Opsamlingsplader, Whatman UniPlate™
PP, 96-brønds, ufarvet, 2000 μl
Catalog # 734-2559
Supplier: Whatman products (Cytiva)
Jump to:
Product Details & Documents
The UNIPLATE™ series includes microplates with a range of well profiles, well volumes and well densities, made from diverse polymer materials. Most UNIPLATE™ microplates conform to the proposed SBS microplate standard and fit most microplate readers and automated plate handling devices.
They are suitable for a wide range of applications, including simple filtrate collection, when used in conjunction with UNIFILTER® microplates, as well as homogeneous assay techniques utilised in HTS.
Specifications
- Farve:ufarvet
- Beskrivelse:PP, 96-brønds
- Brøndvolumen:2000 μl