Orders LinkContactUsLinkComponent
Opsamlingsplader, Whatman UniPlate™
Opsamlingsplader, Whatman UniPlate™
PP, 24-brønds, ufarvet, 10000 μl
Katalog # 734-2558
Opsamlingsplader, Whatman UniPlate™
Katalog # 734-2558
Opsamlingsplader, Whatman UniPlate™
Opsamlingsplader, Whatman UniPlate™
PP, 24-brønds, ufarvet, 10000 μl
Price
4.150,00 kr
Log ind for tilgængelighed
1 * 25 Items
Product Details & Documents
The UNIPLATE™ series includes microplates with a range of well profiles, well volumes and well densities, made from diverse polymer materials. Most UNIPLATE™ microplates conform to the proposed SBS microplate standard and fit most microplate readers and automated plate handling devices.
They are suitable for a wide range of applications, including simple filtrate collection, when used in conjunction with UNIFILTER® microplates, as well as homogeneous assay techniques utilised in HTS.
Specifikationer
  • Farve:
    ufarvet
  • Beskrivelse:
    PP, 24-brønds
  • Brøndvolumen:
    10000 μl