Test Tubes with Screw Cap
10 ml, Yes, Skruelåg, Test tubes, sterile
Katalog # KART5605
New Product
Leverandør: KARTELL
Gå til:
Product Details & Documents
PP test tubes, 10 ml, with self-standing base and tight screw cap yellow colour for a leakproof closure.
Not graduated
Available also in sterile and individually wrapped version
Ø×H: 16×100 mm
Specifikationer
- Kasket inkluderet:Yes
- Lukkestørrelse:Skruelåg
- Kapacitet:10 ml
- Beskrivelse:Test tubes, sterile
- Seq:3