Screw Caps
GL 45
Katalog # HECH41326045
New Product
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
PP, sort.
Very good chemical resistance
- Heat-resistant up to 110 °C
- Only suitable for bottles 215-2814 to 215-2819
Specifikationer
- Gevind:GL 45
- Seq:6