Isolator cleaning tool
Product Details & Documents
Isolator cleaning tool: Mop hardware, replacement head, replacement handle.
Specially engineered to work with isolators and other small areas, this easy to manoeuvre tool offers lightweight, one handed operation. With a low profile, swivel head, the isolator cleaning tool gets into 'hard to reach' places.
Engineered for quick cover changes during operation, the padded head allows for a consistent mop across different surfaces without losing cleaning power. The rounded edges clean the isolator corners and prevent scratches or damage to surfaces.
- Rapid change system allows user to replace covers/pads frequently, adhering to strict cleaning protocols
- Polyester pad conforms to surfaces
- Gets into 'hard to reach' areas with its low profile, swivel head
- 100% polyester covers provide the cleanest substrates available for cleaning isolators
- Offers one handed operation with lightweight handles
Isolator cleaning covers:
Made from 100% polyester or 100% polyester microdenier fabric. This fabric provides enhanced particle lifting, streak-free cleaning, plus enhanced sorption for working with solutions.
Enhanced absorbency and quick-wicking performance
Increased lifting and capturing of particles as small as microbes Streak-free cleaning.
Made from 100% polyester or 100% polyester microdenier fabric. This fabric provides enhanced particle lifting, streak-free cleaning, plus enhanced sorption for working with solutions.
Specifikationer
- Beskrivelse:Sterile Isolator cleaning tool polyester covers and polyester pads
- Seq:8
- Pakket:5 covers and 1 pad/ bag; 25 bags/case