Lovibond® Total Drop Test Kit
Product Details & Documents
Lovibond® offers a wide range of single parameter Drop Test Kits forindividual test requirements. They are for use in various water testing applications including potable water, process water, industrial boilers and cooling systems, swimming sool and waste water treatment.
- Ideal for mobile treatment engineers
- Compact and portable design
- Easy to follow instructions including pictograms
- Complete with all necessary equipment
Drop Tests work by a simple colour change and are derived from laboratory burette methods.
The high quality Lovibond® General Drop Test Kits and reagents are available in a wide range of parameter specific options to suit many analytical requirements. Each self-contained Kit is housed in its own rugged carry case and comes complete with all necessary equipment such as instructions, reagents, syringe and test bottle or test cell for 100 tests.
Derived from laboratory burette methods, the Lovibond® drop test range forms an integral part of the analytical program. The reagents are quick to use and give results fast.
: Includes plastic case, Hardness total buffer TH2, Hardness total indicator TH1P, Hardness LR titrant TH3, Hardness HR titrant TH4, syringe plastic, 20 ml, stirring rod and powder spoon, titration jar with cap, plastic, 60 ml, instruction manual.
Specifikationer
Specifikationer
- Beskrivelse:Sulphite drop test kit
- Måleområde:5 - 150 mg/l Na₂SO₃
- Cat. No.:LOVI56K700360