Tool carts
Tool cart, with 6 drawers, secure lock and cleanroom castors with brakes, 730×470×1000 mm
Catalog # PALBTOOLCART6
New Product
Supplier: Palbam
Jump to:
Product Details & Documents
Tool cart in stainless steel 304 with electropolished finish.
- Five fully extendable drawers
- One lock which secures all the drawers
- Top ledge to prevent tools/parts falling
- Cleanroom castors with brakes
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Beskrivelse:Tool cart, with 6 drawers, secure lock and cleanroom castors with brakes
- Seq:2
- B×D×H:730×470×1000 mm