Combination Blocks
STT-W14-COMBO block, Ø 8,3/13,9/26,2 mm, 6/12/13/25 mm round-bottom tubes, 6/5/3, 48,4 mm
Katalog # VELPA00000511
New Product
Leverandør: VELP SCIENTIFIC
Gå til:
Product Details & Documents
Specifikationer
- Ant. huller:6/5/3
- Beskrivelse:STT-W14-COMBO block
- Seq:1
- Dybde:48,4 mm
- Brøndstørrelse:Ø 8,3/13,9/26,2 mm
- Til:6/12/13/25 mm round-bottom tubes