Blocks for COD
COD-W20-COMBO block, Ø 16,0/22,0 mm, 16/22 mm round-bottom tubes, 18/2, 47/45,5 mm
Katalog # 705-1208
New Product
Leverandør: VELP SCIENTIFIC
Gå til:
Product Details & Documents
Dedicated to COD (Chemical Oxygen Demand) analysis in environmental testing, ensuring accurate digestion of water and wastewater samples under controlled conditions.
Specifikationer
- Ant. huller:18/2
- Beskrivelse:COD-W20-COMBO block
- Seq:2
- Dybde:47/45,5 mm
- Brøndstørrelse:Ø 16,0/22,0 mm
- Til:16/22 mm round-bottom tubes