Accessories for SCAT Level Controls
Holder for disc sensor, GL 45
Catalog # SCTV106723
New Product
Supplier: S.C.A.T.
Jump to:
Product Details & Documents
Necessary and helpful accessories for use with SCAT electronic level controls.
- Switching cables
- Disk sensors
- Signal cables
- Power supply units
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Beskrivelse:Holder for disc sensor, GL 45
- Seq:1
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



