Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Ordreindtastning
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Denmark
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
HARDNESS INDICATOR SOLUTION MANVER 1 * 500 mL
HARDNESS INDICATOR SOLUTION MANVER 1 * 500 mL
Print
HARDNESS INDICATOR SOLUTION MANVER 1 * 500 mL
Catalog # HACH42549-EU
New Product
Supplier:
Hach
HARDNESS INDICATOR SOLUTION MANVER 1 * 500 mL
Catalog # HACH42549-EU
Supplier:
Hach
New Product
Expand image
Return to Top
HARDNESS INDICATOR SOLUTION MANVER 1 * 500 mL
Add to cart
Price
449,00 kr
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Add to cart
Return Policy
Product Details & Documents
Water hardness testing solutions
#: HACH42653
Supplier: Hach
Accessories for test kits, general boiler water, drop count titration
#: HACH2349832
Supplier: Hach
Svovlsyre 0.0175 mol/l (0.035 N) in aqueous solution
#: HACH2349732
Supplier: Hach
Powder pillows, Chloride 2 indicator
#: HACH104399
Supplier: Hach
Reagents for restocking the digital titrator kits
#: 705-0198
Supplier: Hach
Powder pillows, iron
#: HACH85499
Supplier: Hach