Capcell Pak HPLC Columns
Analytisk, SCX UG 80, Rustfrit stål, 80 Å, 5 µm, 4,60 mm, 150 mm
Catalog # PHECCH0-3882
New Product
Supplier: PHENOMENEX
Jump to:
Product Details & Documents
Specifications
- Kolonne form:Analytisk
- Ø indv.:4,60 mm
- Partikelstørrelse:5 µm
- Porestørrelse:80 Å
- Længde:150 mm
- Seq:11
- Kolonnemateriale:Rustfrit stål
- Fase:SCX UG 80
- Cat. No.:PHECCH0-3882
- Endcapped:Ja
- Kulstofbelastning:0%
- pH-område:2,0 - 7,5