Prodigy HPLC Columns, C8
Analytisk, C8 (Octyl), Rustfrit stål, 150 Å, 5 µm, 4,60 mm, 250 mm
Catalog # PHEC00G-3301-E0
New Product
Supplier: PHENOMENEX
Jump to:
Product Details & Documents
Specifications
- Kolonne form:Analytisk
- Ø indv.:4,60 mm
- Partikelstørrelse:5 µm
- Porestørrelse:150 Å
- Længde:250 mm
- Seq:6
- Kolonnemateriale:Rustfrit stål
- Fase:C8 (Octyl)
- Cat. No.:PHEC00G-3301-E0
- Endcapped:Ja
- Kulstofbelastning:12,60%
- pH-område:2,0 - 9,0
- Surface area:310 m²/g