Orders LinkContactUsLinkComponent
Phenol Red indicator
Phenol Red indicator
Bottle
Katalog # APOSOR10146252.5KG
Irriterende
New Product
Leverandør:   Apollo Scientific
Phenol Red indicator
Katalog # APOSOR10146252.5KG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  143-74-8
Phenol Red indicator
Bottle
CAS nummer:  143-74-8
Price
3.750,00 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents
Usage: Organic building block/reagent
Synonymer:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS nummer:143-74-8
  • Formel:C₁₉H₁₄O₅S
  • MDL nummer:MFCD00003552
  • Molecular Weight:354,38 g/mol
  • EINECS:205-609-7
Specifikationer
Specifikationer
  • Pakning:
    Bottle