Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Metanilyellow indicator

Metanilyellow indicator

Bottle
Katalog # APOSOR10137372.5KG
New Product
Leverandør:   Apollo Scientific
Metanilyellow indicator
Katalog # APOSOR10137372.5KG
Leverandør:  Apollo Scientific
New Product
CAS nummer:  587-98-4
Metanilyellow indicator

Bottle

CAS nummer:  587-98-4
Price
2.080,00 kr
Log ind for tilgængelighed
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/ reagent.

Synonymer:  Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
  • CAS nummer:587-98-4
  • Formel:C₁₈H₁₄N₃NaO₃S
  • MDL nummer:MFCD00007486
  • Molecular Weight:375,38 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.