Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Docosanoic acid 85%

Bottle
Katalog # APOSOR1013061-100G
New Product
Leverandør:   Apollo Scientific
Docosanoic acid 85%
Katalog # APOSOR1013061-100G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  112-85-6
Docosanoic acid 85%

Bottle

CAS nummer:  112-85-6
Price
187,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Usage: Organic building block/reagent

Synonymer:  Docosanoic acid
  • CAS nummer:112-85-6
  • Formel:C₂₂H₄₄O₂
  • MDL nummer:MFCD00002807
  • Molecular Weight:340,58 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer