Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Docosanoic acid 85%

Docosanoic acid 85%

Bottle
Katalog # APOSOR1013061-25G
New Product
Leverandør:   Apollo Scientific
Docosanoic acid 85%
Katalog # APOSOR1013061-25G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  112-85-6
Docosanoic acid 85%

Bottle

CAS nummer:  112-85-6
Price
165,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Usage: Organic building block/reagent

Synonymer:  Docosanoic acid
  • CAS nummer:112-85-6
  • Formel:C₂₂H₄₄O₂
  • MDL nummer:MFCD00002807
  • Molecular Weight:340,58 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.