Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Nonanol 98%

2-Nonanol 98%

Bottle
Katalog # APOSOR1030644100ML
New Product
Leverandør:   Apollo Scientific
2-Nonanol 98%
Katalog # APOSOR1030644100ML
Leverandør:  Apollo Scientific
New Product
CAS nummer:  628-99-9
2-Nonanol 98%

Bottle

CAS nummer:  628-99-9
Price
2.280,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

Usage: Organic building block/reagent

Synonymer:  (±)-Heptyl methyl carbinol, (±)-2-Hydroxy nonane, (±)-Methyl heptyl carbinol
  • CAS nummer:628-99-9
  • Formel:C₉H₂₀O
  • Molecular Weight:144,26 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer