Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

2-Nonanol 98%

Bottle
Catalog # APOSOR1030644-25ML
New Product
Supplier:   Apollo Scientific
2-Nonanol 98%
Catalog # APOSOR1030644-25ML
Supplier:  Apollo Scientific
New Product
CAS Number:  628-99-9
2-Nonanol 98%

Bottle

CAS Number:  628-99-9
Price
714,00 kr
Log in for availability
1 * 25 mL
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  (±)-Heptyl methyl carbinol, (±)-2-Hydroxy nonane, (±)-Methyl heptyl carbinol
  • CAS Number:628-99-9
  • Formula:C₉H₂₀O
  • Molecular Weight:144,26 g/mol
Specifications

Specifications

  • Pakning:
    Bottle

Specifications