Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

2-Nonanol 98%

Bottle
Katalog # APOSOR1030644-5ML
New Product
Leverandør:   Apollo Scientific
2-Nonanol 98%
Katalog # APOSOR1030644-5ML
Leverandør:  Apollo Scientific
New Product
CAS nummer:  628-99-9
2-Nonanol 98%

Bottle

CAS nummer:  628-99-9
Price
231,00 kr
Log ind for tilgængelighed
1 * 5 mL
Product Details & Documents

Usage: Organic building block/reagent

Synonymer:  (±)-Heptyl methyl carbinol, (±)-2-Hydroxy nonane, (±)-Methyl heptyl carbinol
  • CAS nummer:628-99-9
  • Formel:C₉H₂₀O
  • Molecular Weight:144,26 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.