Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Finasteride 98%

Finasteride 98%

Bottle
Katalog # APOSBI12115-100G
New Product
Leverandør:   Apollo Scientific
Finasteride 98%
Katalog # APOSBI12115-100G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  98319-26-7
Finasteride 98%

Bottle

CAS nummer:  98319-26-7
Price
15.190,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonymer:  N-tert-Butyl-3-oxo-4-aza-5alpha-androst-1-en-17beta-carboxamide
  • CAS nummer:98319-26-7
  • Formel:C₂₃H₃₆N₂O₂
  • MDL nummer:MFCD00869737
  • Molecular Weight:372,549 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.