Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Metanilyellow indicator

Metanilyellow indicator

Bottle
Katalog # APOSOR1013737-5G
New Product
Leverandør:   Apollo Scientific
Metanilyellow indicator
Katalog # APOSOR1013737-5G
Leverandør:  Apollo Scientific
New Product
CAS nummer:  587-98-4
Metanilyellow indicator

Bottle

CAS nummer:  587-98-4
Price
165,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents

Usage: Organic building block/ reagent.

Synonymer:  Acid Yellow 36, 3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt
  • CAS nummer:587-98-4
  • Formel:C₁₈H₁₄N₃NaO₃S
  • MDL nummer:MFCD00007486
  • Molecular Weight:375,38 g/mol
Specifikationer

Specifikationer

  • Pakning:
    Bottle

Specifikationer