Gemfibrozil ≥98%
Bottle
Catalog # APOSBUP14848-200MG
New Product
Supplier: Apollo Scientific
Gemfibrozil ≥98%
Catalog # APOSBUP14848-200MG
Supplier: Apollo Scientific
New Product
CAS Number: 25812-30-0
Gå til:
Product Details & Documents
Usage: Biochemical/life science applications
Synonymer: CI-719
- CAS nummer:25812-30-0
- Formel:C₁₅H₂₂O₃
- Molecular Weight:250,33 g/mol
Specifikationer
Specifikationer
- Pakning:Bottle
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



